Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Rabgef1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199059.1 | NP_001185988.1 | 4 | 557 | ||
NM_019983.2 | NP_064367.1 | 3 | 702 | |||
GenBank mRNA | AF093590.1 | - | 4 | 533 | ||
AK044568.1 | - | 3 | 722 | |||
AK146734.1 | - | 4 | 507 | |||
BC018229.1 | - | 3 | 702 | 4507477 |