Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rec8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020002.3 | NP_064386.2 | 3 | 337 | ||
GenBank mRNA | AF262055.1 | - | 3 | 267 | ||
AK077167.1 | - | 3 | 272 | |||
AK146832.1 | - | 3 | 323 | |||
AU080225.1 | - | 3 | 338 | |||
BC052155.1 | - | 3 | 333 | 6335959 |