Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldh9a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019993.3 | NP_064377.2 | 3 | 617 | ||
GenBank mRNA | AB296386.1 | - | 3 | 562 | ||
AF170919.1 | - | 3 | 362 | |||
AK132336.1 | - | 3 | 402 | |||
AK149589.1 | - | 3 | 403 | |||
AK150093.1 | - | 3 | 360 | |||
AK154914.1 | - | 3 | 601 | |||
AK162354.1 | - | 3 | 401 | |||
AK165730.1 | - | 3 | 617 | |||
AK168261.1 | - | 3 | 595 | |||
AK168309.1 | - | 3 | 595 | |||
AK168878.1 | - | 3 | 566 | |||
BC003297.1 | - | 3 | 394 | 3583976 |