Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem9b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020050.1 | NP_064434.1 | 4 | 450 | ||
GenBank mRNA | AK007430.1 | - | 4 | 418 | ||
AK009147.1 | - | 4 | 385 | |||
AK049882.1 | - | 4 | 537 | |||
AK077253.1 | - | 4 | 406 | |||
AK147127.1 | - | 4 | 466 | |||
AK201035.1 | - | 3 | 264 | |||
BC002208.1 | - | 4 | 434 | 3490055 |