Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LCP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005565.3 | NP_005556.1 | 7 | 721 | ||
GenBank mRNA | AK292890.1 | - | 7 | 682 | ||
AK300000.1 | - | 5 | 565 | |||
BC016618.1 | - | 7 | 658 | 4476274 | ||
BT007273.1 | - | 7 | 514 | |||
FN432830.1 | - | 8 | 594 | |||
U20158.1 | - | 7 | 769 |