Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020603.2 | NP_065628.1 | 15 | 1831 | ||
GenBank mRNA | AK009209.1 | - | 3 | 247 | ||
AK145068.1 | - | 15 | 1886 | |||
AK216336.1 | - | 1 | 76 | |||
BC026431.1 | - | 12 | 1455 | |||
BC046977.1 | - | 15 | 1831 | 6315172 | ||
KJ650211.1 | - | 15 | 1816 | |||
KJ663716.1 | - | 15 | 1684 |