Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Wdr46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020603.2 | NP_065628.1 | 11 | 1277 | ||
GenBank mRNA | AK145068.1 | - | 11 | 1332 | ||
BC013527.1 | - | 11 | 1320 | |||
BC026431.1 | - | 8 | 901 | |||
BC046977.1 | - | 11 | 1277 | 6315172 | ||
KJ650211.1 | - | 11 | 1262 | |||
KJ663716.1 | - | 11 | 1130 |