Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023785.2 | NP_076274.1 | 2 | 244 | ||
GenBank mRNA | AB042817.1 | - | 2 | 253 | ||
AF219112.1 | - | 2 | 244 | |||
AF278700.1 | - | 2 | 244 | |||
AK010273.1 | - | 2 | 245 | |||
AK157077.1 | - | 2 | 244 | |||
AK157141.1 | - | 2 | 245 | |||
AK158061.1 | - | 2 | 242 | |||
AK171720.1 | - | 2 | 245 | |||
BC127043.1 | - | 2 | 211 | 40053104 | ||
BC127044.1 | - | 2 | 211 | 40053105 |