Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Atp5j2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020582.2 | NP_065607.1 | 3 | 215 | ||
GenBank mRNA | AB030192.1 | - | 3 | 216 | ||
AK002519.1 | - | 3 | 165 | |||
AK003235.1 | - | 3 | 153 | |||
AK003817.1 | - | 3 | 165 | |||
AK013130.1 | - | 3 | 165 | |||
AK151737.1 | - | 3 | 164 | |||
AK168095.1 | - | 3 | 165 | |||
BC029226.1 | - | 2 | 155 | 2651401 |