Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020593.2 | NP_065618.1 | 8, 9 | 1004 | ||
NM_212433.1 | NP_997598.1 | 8, 9 | 1004 | |||
GenBank mRNA | AF233226.1 | - | 8, 9 | 943 | ||
AK004544.2 | - | 8, 9 | 956 | |||
AK006392.1 | - | 1, 2 | 116 | |||
AK009024.1 | - | 8, 9 | 943 | |||
AK019550.1 | - | 8, 9 | 944 | |||
AK028048.1 | - | 8, 9 | 966 | |||
AK029594.1 | - | 1, 2 | 123 | |||
AK031185.1 | - | 8, 9 | 941 | |||
AK031428.1 | - | 8, 9 | 964 | |||
AK049870.1 | - | 8, 9 | 962 | |||
AK050195.1 | - | 8, 9 | 956 | |||
AK085686.1 | - | 8, 9 | 959 | |||
AK151130.1 | - | 8, 9 | 955 | |||
AK151857.1 | - | 8, 9 | 955 | |||
AK152780.1 | - | 8, 9 | 955 | |||
AK167075.1 | - | 8, 9 | 942 | |||
AK176959.1 | - | 1, 2 | 39 | |||
BC010212.1 | - | 8, 9 | 905 | |||
BC050758.1 | - | 8, 9 | 957 | |||
BC058253.1 | - | 8, 9 | 961 | 6465107 |