Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020593.2 | NP_065618.1 | 7 | 809 | ||
NM_212433.1 | NP_997598.1 | 7 | 809 | |||
GenBank mRNA | AF233226.1 | - | 7 | 748 | ||
AK004544.2 | - | 7 | 761 | |||
AK009024.1 | - | 7 | 748 | |||
AK019550.1 | - | 7 | 749 | |||
AK028048.1 | - | 7 | 771 | |||
AK031185.1 | - | 7 | 748 | |||
AK031428.1 | - | 7 | 769 | |||
AK049870.1 | - | 7 | 767 | |||
AK050195.1 | - | 7 | 761 | |||
AK085686.1 | - | 7 | 764 | |||
AK150097.1 | - | 7 | 761 | |||
AK151130.1 | - | 7 | 760 | |||
AK151857.1 | - | 7 | 760 | |||
AK152780.1 | - | 7 | 760 | |||
AK167075.1 | - | 7 | 747 | |||
BC010212.1 | - | 7 | 710 | |||
BC050758.1 | - | 7 | 762 | |||
BC058253.1 | - | 7 | 766 | 6465107 |