Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LETM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF061025.1 | - | 12 | 2095 | ||
BC014500.1 | - | 12 | 1995 | 4860194 | ||
BC021208.1 | - | 12 | 2020 | 4126510 |