Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Angptl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020581.2 | NP_065606.2 | 7 | 1331 | ||
GenBank mRNA | AB054540.1 | - | 7 | 1217 | ||
AF123261.1 | - | 7 | 1262 | |||
AF169313.1 | - | 7 | 1194 | |||
AF278699.1 | - | 7 | 1266 | |||
AK009161.1 | - | 7 | 1037 | |||
AK014564.1 | - | 1332 | ||||
AK032491.1 | - | 7 | 1299 | |||
AK132761.1 | - | 7 | 1300 | |||
AK134419.1 | - | 7 | 1296 | |||
BC006611.1 | - | 7 | 1283 | 3966729 | ||
BC021343.1 | - | 7 | 1297 | 4194173 | ||
BC025797.1 | - | 7 | 1267 | 5137159 |