Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc42se1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038708.3 | NP_001033797.1 | 5 | 661 | ||
NM_172395.3 | NP_765983.1 | 5 | 1031 | |||
GenBank mRNA | AK004859.1 | - | 5 | 676 | ||
AK008084.1 | - | 4 | 702 | |||
AK019052.1 | - | 5 | 673 | |||
AK028573.1 | - | 5 | 673 | |||
AK213146.1 | - | 3 | 247 | |||
BC029194.1 | - | 5 | 649 | |||
BC060964.1 | - | 5 | 653 | 6743495 |