Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hs1bp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021429.3 | NP_067404.2 | 7 | 1162 | ||
GenBank mRNA | AJ132192.1 | - | 7 | 1156 | ||
AK040654.1 | - | 7 | 1163 | |||
AK043263.1 | - | 3 | 412 | |||
AK084802.1 | - | 7 | 1153 | |||
AK154696.1 | - | 7 | 1158 | |||
AK170838.1 | - | 7 | 1149 | |||
BC095977.1 | - | 7 | 1159 |