Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr180 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021434.5 | NP_067409.2 | 8 | 1214 | ||
GenBank mRNA | AB041545.1 | - | 8 | 1206 | ||
AK040165.1 | - | 2 | 110 | |||
AK053422.1 | - | 8 | 1184 | |||
AK169908.1 | - | 8 | 1213 | |||
AK170316.1 | - | 8 | 1179 | |||
BC055726.1 | - | 8 | 1189 | |||
BC069981.1 | - | 8 | 1170 | 6336270 |