Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhot1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163354.1 | NP_001156826.1 | 16 | 1597 | ||
NM_001163355.1 | NP_001156827.1 | 16 | 1597 | |||
NM_021536.7 | NP_067511.4 | 16 | 1597 | |||
GenBank mRNA | AF244542.1 | - | 3 | 231 | ||
AK019059.1 | - | 16 | 1557 | |||
AK028597.1 | - | 16 | 1540 | |||
AK031139.1 | - | 14 | 1259 | |||
AK032096.1 | - | 16 | 1570 | |||
AK041119.1 | - | 16 | 1533 | |||
AK043323.1 | - | 7 | 654 | |||
AK054472.1 | - | 16 | 1568 | |||
AK078955.1 | - | 16 | 1540 | |||
AK090071.1 | - | 9 | 923 | |||
AK138830.1 | - | 16 | 1538 | |||
AK141621.1 | - | 16 | 1424 | |||
AK147749.1 | - | 16 | 1576 | |||
AK147801.1 | - | 16 | 1602 | |||
AK167502.1 | - | 16 | 1597 | |||
AK169811.1 | - | 15 | 2254 | |||
BC006918.1 | - | 7 | 688 | |||
BC025021.1 | - | 2 | 710 | |||
BC044301.1 | - | 5 | 436 | |||
BC046785.1 | - | 16 | 1580 | 6416127 | ||
BC052882.1 | - | 6 | 541 | |||
BC058350.1 | - | 16 | 1512 | 6815832 |