Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nsa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021552.5 | NP_067527.3 | 2 | 315 | ||
GenBank mRNA | AF238866.1 | - | 2 | 242 | ||
AK002474.1 | - | 1 | 166 | |||
AK009136.1 | - | 1 | 244 | |||
AK012832.1 | - | 2 | 256 | |||
AK014275.1 | - | 2 | 292 | |||
AK017605.1 | - | 2 | 256 | |||
AK019379.1 | - | 2 | 279 | |||
AK077696.1 | - | 257 | ||||
AK078457.1 | - | 2 | 268 | |||
BC086776.1 | - | 1 | 111 | |||
BC108365.1 | - | 2 | 241 | 4219428 | ||
BC116384.1 | - | 2 | 256 | |||
BC126976.1 | - | 2 | 255 | 40101726 | ||
BC172160.1 | - | 2 | 229 | |||
CJ132894.1 | - | 2 | 316 |