Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nsa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021552.5 | NP_067527.3 | 2 | 344 | ||
GenBank mRNA | AK002474.1 | - | 1 | 195 | ||
AK009136.1 | - | 1 | 273 | |||
AK012832.1 | - | 2 | 285 | |||
AK014275.1 | - | 2 | 321 | |||
AK017605.1 | - | 2 | 285 | |||
AK019991.1 | - | 2 | 291 | |||
AK077696.1 | - | 286 | ||||
AK078457.1 | - | 2 | 297 | |||
BC086776.1 | - | 1 | 140 | |||
BC108365.1 | - | 2 | 270 | 4219428 | ||
BC116384.1 | - | 2 | 285 | |||
BC126976.1 | - | 2 | 284 | 40101726 | ||
BC172160.1 | - | 2 | 258 | |||
CJ132894.1 | - | 2 | 345 |