Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hgh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021555.2 | NP_067530.2 | 2 | 762 | ||
GenBank mRNA | AB041552.1 | - | 2 | 762 | ||
AK085815.1 | - | 2 | 756 | |||
AK181958.1 | - | 1 | 30 | |||
AK194217.1 | - | 2 | 124 | |||
BC061251.1 | - | 2 | 726 | 30249656 |