Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Nek6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159631.1 | NP_001153103.1 | 4, 5 | 435 | ||
NM_021606.3 | NP_067619.1 | 4, 5 | 428 | |||
GenBank mRNA | AF218847.1 | - | 4, 5 | 430 | ||
AK004925.1 | - | 4, 5 | 405 | |||
AK011519.1 | - | 4, 5 | 414 | |||
AK018168.1 | - | 4, 5 | 418 | |||
AK029266.1 | - | 4, 5 | 417 | |||
AK075717.1 | - | 4, 5 | 402 | |||
AK075836.1 | - | 4, 5 | 414 | |||
AK086700.1 | - | 433 | ||||
AK089919.1 | - | 4, 5 | 389 | |||
AK170757.1 | - | 4, 5 | 435 | |||
BC019524.1 | - | 4, 5 | 497 | 4209097 |