Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncstn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021607.3 | NP_067620.3 | 9 | 1091 | ||
GenBank mRNA | AF240469.1 | - | 9 | 1041 | ||
AK033314.1 | - | 9 | 1034 | |||
AK129102.1 | - | 9 | 1091 | |||
AK170173.1 | - | 9 | 1037 | |||
AK170244.1 | - | 9 | 1056 | |||
AK170369.1 | - | 9 | 1080 | |||
AK172579.1 | - | 9 | 1036 | |||
BC019998.1 | - | 9 | 1036 | 4458780 |