Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cables1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146287.1 | NP_001139759.1 | 6 | 1653 | ||
NM_022021.2 | NP_071304.2 | 5 | 1575 | |||
GenBank mRNA | AF133208.1 | - | 5 | 1574 | ||
AF328140.1 | - | 5 | 1260 | |||
AK156273.1 | - | 6 | 1709 | |||
AK158396.1 | - | 6 | 1383 | |||
AK187559.1 | - | 3 | 249 | |||
BC043661.1 | - | 5 | 777 | |||
BC145142.1 | - | 5 | 1559 | 9053693 | ||
BC150714.1 | - | 5 | 1559 | 9087625 | ||
BQ917292.1 | - | 3 | 260 |