Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LLGL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015002.1 | NP_001015002.1 | 7 | 794 | ||
NM_001031803.1 | NP_001026973.1 | 7 | 794 | |||
NM_004524.2 | NP_004515.2 | 7 | 794 | |||
XM_011524801.1 | XP_011523103.1 | 7 | 777 | |||
XM_011524802.1 | XP_011523104.1 | 7 | 765 | |||
XM_011524807.2 | XP_011523109.1 | 7 | 882 | |||
XM_017024625.1 | XP_016880114.1 | 7 | 765 | |||
XM_017024626.1 | XP_016880115.1 | 7 | 765 | |||
XM_017024627.1 | XP_016880116.1 | 7 | 765 | |||
XM_017024628.1 | XP_016880117.1 | 7 | 765 | |||
XM_017024629.1 | XP_016880118.1 | 7 | 765 | |||
XM_017024630.1 | XP_016880119.1 | 7 | 765 | |||
XM_017024631.1 | XP_016880120.1 | 7 | 765 | |||
GenBank mRNA | AK125563.1 | - | 1 | 846 | ||
AK126687.1 | - | 4 | 817 | |||
AK315126.1 | - | 7 | 744 | |||
BC006503.2 | - | 8 | 852 | |||
BC010879.1 | - | 7 | 795 | |||
BC064994.1 | - | 7 | 767 | 5575350 | ||
X87342.1 | - | 7 | 770 |