Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001104529.1 | NP_001097999.1 | 2 | 1138 | ||
NM_001271766.1 | NP_001258695.1 | 2 | 1077 | |||
NM_022320.4 | NP_071715.3 | 2 | 1374 | |||
GenBank mRNA | AF200349.1 | - | 2 | 1059 | ||
AK034870.1 | - | 2 | 1158 | |||
AK036503.1 | - | 2 | 1136 | |||
AK154203.1 | - | 2 | 1062 | |||
AK160972.1 | - | 2 | 550 | |||
AK170995.1 | - | 2 | 1078 | |||
BC027429.1 | - | 2 | 1011 | 3669814 |