Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Parvg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162500.1 | NP_001155972.1 | 9 | 771 | ||
NM_022321.4 | NP_071716.3 | 9 | 726 | |||
GenBank mRNA | AF312712.1 | - | 9 | 684 | ||
AK028329.1 | - | 9 | 727 | |||
AK037365.1 | - | 9 | 699 | |||
AK040914.1 | - | 9 | 657 | |||
AK079881.1 | - | 9 | 652 | |||
AK088419.1 | - | 9 | 676 | |||
BC011200.1 | - | 9 | 680 | 4190510 |