Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnmd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022322.2 | NP_071717.1 | 4, 5 | 630 | ||
GenBank mRNA | AB055422.1 | - | 4, 5 | 437 | ||
AF191768.1 | - | 4, 5 | 576 | |||
AF219993.1 | - | 4, 5 | 503 | |||
AF291655.1 | - | 4, 5 | 655 | |||
AK003748.1 | - | 4, 5 | 514 | |||
AK014761.1 | - | 4, 5 | 469 | |||
BC006919.1 | - | 4, 5 | 439 | 3601223 | ||
BC049944.1 | - | 4, 5 | 630 | 2811628 |