Gene Symbol: Stk32b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022416.2 | NP_071861.1 | 12 | 1560 | ||
GenBank mRNA | AJ250840.1 | - | 12 | 1638 | ||
AK082468.1 | - | 12 | 1561 | |||
BC052404.1 | - | 12 | 1470 | 5715729 | ||
BC056396.1 | - | 12 | 1546 | 6841272 | ||
BC058412.1 | - | 12 | 1470 | 5715729 |