Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Fgf23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022657.4 | NP_073148.1 | 3 | 1305 | ||
GenBank mRNA | AF263536.1 | - | 3 | 1233 | ||
AK155687.1 | - | 3 | 1305 | |||
BC120605.1 | - | 3 | 1179 | 40129528 |