Search Thermo Fisher Scientific
- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Search Thermo Fisher Scientific
Gene Symbol: Mrps14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025474.3 | NP_079750.1 | 3 | 253 | ||
GenBank mRNA | AK007673.1 | - | 3 | 266 | ||
AK010242.1 | - | 3 | 259 | |||
AK083352.1 | - | 3 | 253 | |||
AK144195.1 | - | 260 | ||||
BC044057.1 | - | 3 | 247 | 6489566 |