Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Pes1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022889.3 | NP_075027.1 | 5 | 539 | ||
GenBank mRNA | AF289539.1 | - | 5 | 499 | ||
AK029075.1 | - | 5 | 539 | |||
AK088831.1 | - | 5 | 517 | |||
BC004844.1 | - | 5 | 524 | 2649598 | ||
BC011142.1 | - | 5 | 522 | 4207164 |