Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prrg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022999.1 | NP_075375.1 | 2 | 201 | ||
GenBank mRNA | AK005453.1 | - | 3 | 245 | ||
BC025098.1 | - | 2 | 201 | 4159863 |