Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000015.2 | NP_000006.2 | 2 | 274 | ||
XM_017012938.1 | XP_016868427.1 | 2 | 625 | |||
GenBank mRNA | AJ581144.1 | - | 2 | 205 | ||
AJ581145.1 | - | 1 | 234 | |||
BC015878.1 | - | 2 | 236 | 4716636 | ||
BC067218.1 | - | 2 | 271 | 4722596 | ||
CR407631.1 | - | 1 | 167 | |||
D90040.1 | - | 2 | 208 | |||
D90042.1 | - | 2 | 274 |