Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sdhaf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025333.4 | NP_079609.2 | 3 | 455 | ||
GenBank mRNA | AK013454.1 | - | 3 | 360 | ||
AK075594.1 | - | 3 | 360 | |||
AK160175.1 | - | 1 | 88 | |||
AK160957.1 | - | 3 | 362 | |||
AK167228.1 | - | 3 | 391 | |||
BC029630.1 | - | 2 | 174 | |||
BC038924.1 | - | 2 | 423 | |||
BC048913.1 | - | 3 | 188 | |||
BC058955.1 | - | 3 | 404 | |||
BC060090.1 | - | 3 | 404 | 5698361 | ||
BC085277.1 | - | 3 | 357 | 30441476 |