Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chchd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167736.1 | NP_001161208.1 | 3 | 474 | ||
NM_025351.3 | NP_079627.3 | 3 | 474 | |||
GenBank mRNA | AK002980.1 | - | 3 | 458 | ||
AK005696.1 | - | 3 | 406 | |||
AK006192.1 | - | 3 | 410 | |||
AK015314.1 | - | 3 | 437 | |||
AK087642.1 | - | 3 | 383 | |||
AK144093.1 | - | 3 | 380 | |||
AK167314.1 | - | 3 | 427 | |||
BC011331.1 | - | 3 | 437 | 3153325 | ||
BC052929.1 | - | 3 | 433 | 1366861 | ||
BQ177996.1 | - | 3 | 435 | |||
EF410100.1 | - | 534 |