Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufa9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025358.3 | NP_079634.2 | 5 | 492 | ||
GenBank mRNA | AK003137.1 | - | 5 | 474 | ||
AK003214.1 | - | 5 | 465 | |||
AK012366.1 | - | 5 | 472 | |||
BC005760.1 | - | 4 | 373 | |||
BC058378.1 | - | 5 | 460 | 6820787 |