Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Fkbp11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024169.3 | NP_077131.2 | 6 | 638 | ||
GenBank mRNA | AK003331.1 | - | 6 | 639 | ||
AK007813.1 | - | 6 | 602 | |||
AK019132.1 | - | 6 | 580 | |||
BC002311.1 | - | 7 | 588 | |||
BC022900.1 | - | 4 | 1207 | |||
BC037596.1 | - | 6 | 594 | 5250344 |