Search Thermo Fisher Scientific
Gene Symbol: Fkbp11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024169.3 | NP_077131.2 | 3 | 336 | ||
GenBank mRNA | AK003331.1 | - | 3 | 337 | ||
AK007813.1 | - | 3 | 300 | |||
AK019132.1 | - | 3 | 278 | |||
BC002311.1 | - | 3 | 290 | |||
BC021345.1 | - | 1 | 871 | |||
BC022900.1 | - | 3 | 290 | |||
BC037596.1 | - | 3 | 292 | 5250344 |