Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acyp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025421.2 | NP_079697.1 | 2 | 178 | ||
GenBank mRNA | AK004176.1 | - | 2 | 178 | ||
AK007244.1 | - | 2 | 160 | |||
AK018832.1 | - | 2 | 128 | |||
AK027969.1 | - | 2 | 162 | |||
BC061088.1 | - | 2 | 149 | 6773293 | ||
BC096417.1 | - | 2 | 151 | |||
BC116781.1 | - | 2 | 131 | 40126100 | ||
BC116783.1 | - | 2 | 131 | 40126102 |