Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004525.2 | NP_004516.2 | 69 | 12880 | ||
XM_011511183.2 | XP_011509485.1 | 68 | 12752 | |||
XM_011511184.2 | XP_011509486.1 | 54 | 10393 | |||
GenBank mRNA | AK302843.1 | - | 20 | 2779 | ||
AY265357.1 | - | 69 | 12734 | |||
AY265358.1 | - | 69 | 12731 | |||
S73145.1 | - | 3 | 253 | |||
U33837.1 | - | 69 | 12771 |