Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thoc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285780.1 | NP_001272709.1 | 4 | 400 | ||
NM_025435.3 | NP_079711.1 | 3 | 322 | |||
GenBank mRNA | AK005176.1 | - | 3 | 249 | ||
AK007440.1 | - | 3 | 228 | |||
AK008899.1 | - | 4 | 305 | |||
AK010958.1 | - | 3 | 263 | |||
AK013319.1 | - | 3 | 245 | |||
AK045545.1 | - | 1 | 2233 | |||
AK143106.1 | - | 2 | 1222 | |||
AK147003.1 | - | 4 | 303 | |||
BC027113.1 | - | 2 | 1270 | |||
BC042209.1 | - | 3 | 249 | |||
BC054419.1 | - | 3 | 282 | 4010343 | ||
BC116919.1 | - | 3 | 287 | 40126238 | ||
BC116945.1 | - | 3 | 287 | 40126264 | ||
CK329337.1 | - | 3 | 323 |