Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Defa22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207658.4 | NP_997541.2 | 2 | 236 | ||
GenBank mRNA | AK008589.1 | - | 2 | 237 | ||
AY746427.1 | - | 2 | 193 |
Gene Symbol: Defa21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183253.3 | NP_899076.1 | 2 | 236 | ||
GenBank mRNA | AK008266.1 | - | 2 | 237 | ||
AY746426.1 | - | 2 | 193 |