Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Senp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK010813.1 | - | 5 | 462 | ||
AK012740.1 | - | 6 | 554 | |||
AK013090.1 | - | 6 | 554 | |||
AK020068.1 | - | 5 | 457 | |||
AK083086.1 | - | 6 | 539 | |||
AK085060.1 | - | 6 | 553 | |||
AK122542.1 | - | 6 | 549 | |||
AK164354.1 | - | 6 | 538 | |||
BC058593.1 | - | 6 | 496 | 5364345 | ||
BY016675.1 | - | 4 | 330 | |||
CF583861.1 | - | 7 | 752 |