Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Atp6v1c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025494.3 | NP_079770.2 | 2 | 268 | ||
GenBank mRNA | AB088407.1 | - | 2 | 215 | ||
AK006323.1 | - | 2 | 219 | |||
AK075779.1 | - | 2 | 240 | |||
AK129007.1 | - | 2 | 228 | |||
AK151237.1 | - | 2 | 229 | |||
BB869068.1 | - | 2 | 269 | |||
BC010217.1 | - | 2 | 216 | 3498388 | ||
U13839.1 | - | 2 | 211 |