Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc90b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162918.1 | NP_001156390.1 | 3 | 676 | ||
NM_025515.3 | NP_079791.2 | 3 | 676 | |||
GenBank mRNA | AK009370.1 | - | 3 | 297 | ||
AK011631.1 | - | 2 | 279 | |||
AK084673.1 | - | 3 | 311 | |||
AK160230.1 | - | 3 | 332 | |||
BC034878.1 | - | 3 | 606 | 3468518 | ||
BC115973.1 | - | 3 | 297 | 40100643 |