Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc90b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025515.3 | NP_079791.2 | 4 | 782 | ||
GenBank mRNA | AK009370.1 | - | 4 | 403 | ||
AK011631.1 | - | 3 | 385 | |||
AK084673.1 | - | 4 | 417 | |||
AK160230.1 | - | 4 | 438 | |||
BC115973.1 | - | 4 | 403 | 40100643 |