Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2310011J03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025521.3 | NP_079797.1 | 2 | 1181 | ||
GenBank mRNA | AK009305.1 | - | 2 | 1164 | ||
AK020401.1 | - | 2 | 1171 | |||
AK046452.1 | - | 2 | 1142 | |||
AK140230.1 | - | 2 | 1142 | |||
AK180888.1 | - | 1 | 403 | |||
BC023429.1 | - | 2 | 1148 | 5042442 | ||
BY596174.1 | - | 1 | 13 |