Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other LTBP1 SIRNA Assays ›
Gene Symbol:
LTBP1
Gene Name:
latent transforming growth factor beta binding protein 1
Gene Aliases:
-
Chromosome Location:
Chr.2: 32947129 - 33399509 on Build GRCh38
UniGene ID:
Hs.619315
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s8318
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: LTBP1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000627.3
NP_000618.3
6
1086
NM_001166264.1
NP_001159736.1
6
1086
NM_001166265.1
NP_001159737.1
6
1086
NM_001166266.1
NP_001159738.1
6
1086
NM_206943.2
NP_996826.2
10
1946
XM_005264317.3
XP_005264374.1
10
1948
XM_005264318.3
XP_005264375.1
10
1948
XM_011532853.2
XP_011531155.1
10
2119
XM_011532855.2
XP_011531157.1
10
1948
XM_011532856.2
XP_011531158.1
10
1948
XM_011532857.2
XP_011531159.1
10
1948
XM_011532858.2
XP_011531160.1
10
1948
XM_011532859.2
XP_011531161.1
10
1948
XM_011532860.2
XP_011531162.1
9
1780
XM_011532861.2
XP_011531163.1
10
1948
XM_011532862.2
XP_011531164.1
10
1948
XM_017004108.1
XP_016859597.1
10
1948
XM_017004109.1
XP_016859598.1
10
1948
XM_017004110.1
XP_016859599.1
10
1948
GenBank mRNA
AB208801.1
-
6
1044
AF489528.1
-
6
945
AK299912.1
-
1
43
BC130289.1
-
6
1026
40146320
BC144127.1
-
6
1026
BC144128.1
-
6
1026
9052648
CF995689.1
-
1
71
M34057.1
-
6
1035
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_206958, NM_019919, BC094612, AF022889, AK036047.
Panther Classification:
Molecular Function -
extracellular matrix structural protein
Gene Ontology Categories:
Function(s)
Process(es)
ventricular septum development
transmembrane receptor protein serine/threonine kinase signaling pathway
sequestering of TGFbeta in extracellular matrix
aorta development
coronary vasculature development
transforming growth factor beta-activated receptor activity
calcium ion binding
protein binding
transforming growth factor beta binding
microfibril binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-7cf5v:80/10.239.163.60:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0