Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025567.2 | NP_079843.1 | 4 | 555 | ||
GenBank mRNA | AK002815.1 | - | 4 | 533 | ||
AK011288.1 | - | 4 | 556 | |||
AK151299.1 | - | 4 | 546 | |||
AK170272.1 | - | 3 | 1155 | |||
AK186904.1 | - | 2 | 118 | |||
AK192516.1 | - | 4 | 412 | |||
BC005620.1 | - | 4 | 536 | 3711323 | ||
BC082790.1 | - | 4 | 493 |