Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnpy4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178612.4 | NP_848727.1 | 3 | 364 | ||
GenBank mRNA | AK011474.1 | - | 3 | 365 | ||
AK012968.1 | - | 3 | 346 | |||
AK033219.1 | - | 3 | 366 | |||
AK051562.1 | - | 3 | 366 | |||
BC086647.1 | - | 3 | 329 | 6818372 | ||
BC094672.1 | - | 3 | 471 | 30633103 |